siRNA Details
| Gene Symbol: Trim66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191839.1 | NP_001178768.1 | 10 | 2060 | ||
| XM_006229983.2 | XP_006230045.1 | 11 | 3257 | |||
| GenBank mRNA | BC168212.1 | - | 11 | 1947 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim66| Gene Symbol: Trim66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191839.1 | NP_001178768.1 | 10 | 2060 | ||
| XM_006229983.2 | XP_006230045.1 | 11 | 3257 | |||
| GenBank mRNA | BC168212.1 | - | 11 | 1947 | ||