siRNA Details
Gene Symbol: Cyp2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108499.1 | NP_001101969.1 | 5 | 804 | ||
XM_006230081.2 | XP_006230143.1 | 5 | 1815 | |||
XM_006230084.2 | XP_006230146.1 | 5 | 1594 | |||
GenBank mRNA | BC168986.1 | - | 4 | 1222 | ||
FQ219437.1 | - | 4 | 1118 |
