siRNA Details
| Gene Symbol: Ap5b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271032.1 | NP_001257961.1 | 3 | 359 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap5b1| Gene Symbol: Ap5b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271032.1 | NP_001257961.1 | 3 | 359 | ||