siRNA Details
Gene Symbol: Dpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108516.1 | NP_001101986.1 | 2 | 194 | ||
XM_006230850.1 | XP_006230912.1 | 2 | 219 | |||
GenBank mRNA | BC166788.1 | - | 2 | 141 |
