siRNA Details
| Gene Symbol: Dnajc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013196.1 | NP_001013214.1 | 4 | 476 | ||
| XM_006230859.2 | XP_006230921.1 | 4 | 471 | |||
| XM_006230860.2 | XP_006230922.1 | 4 | 433 | |||
| XM_006230861.2 | XP_006230923.1 | 3 | 631 | |||
| XM_006230862.2 | XP_006230924.1 | 3 | 651 | |||
| XM_008760159.1 | XP_008758381.1 | 4 | 470 | |||
| GenBank mRNA | BC088201.1 | - | 4 | 476 | 7378025 | |

Dnajc4
Genomic Map