siRNA Details
| Gene Symbol: Bscl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012171.1 | NP_001012171.1 | 2, 3 | 609 | ||
| GenBank mRNA | BC089942.1 | - | 2, 3 | 609 | 7311394 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bscl2| Gene Symbol: Bscl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012171.1 | NP_001012171.1 | 2, 3 | 609 | ||
| GenBank mRNA | BC089942.1 | - | 2, 3 | 609 | 7311394 | |