siRNA Details
| Gene Symbol: Rfx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012172.1 | NP_001012172.1 | 9 | 1019 | ||
| XM_006231233.2 | XP_006231295.1 | 10 | 1224 | |||
| XM_006231235.2 | XP_006231297.1 | 9 | 1183 | |||
| XM_008760340.1 | XP_008758562.1 | 8 | 992 | |||
| XM_008760341.1 | XP_008758563.1 | 9 | 1067 | |||
| GenBank mRNA | BC085953.1 | - | 9 | 1019 | 7134397 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001166414, NM_011265, BC017598, AK041120, AK132267, and others.

Rfx3
Genomic Map