siRNA Details
| Gene Symbol: Rfx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012172.1 | NP_001012172.1 | 4 | 459 | ||
| XM_006231233.2 | XP_006231295.1 | 5 | 622 | |||
| XM_006231235.2 | XP_006231297.1 | 4 | 581 | |||
| XM_008760340.1 | XP_008758562.1 | 4 | 465 | |||
| XM_008760341.1 | XP_008758563.1 | 4 | 465 | |||
| GenBank mRNA | BC085953.1 | - | 4 | 459 | 7134397 | |

Rfx3
Genomic Map