siRNA Details
| Gene Symbol: Btrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007148.2 | NP_001007149.2 | 2 | 161 | ||
| XM_006231529.2 | XP_006231591.1 | 2 | 744 | |||
| XM_006231531.2 | XP_006231593.1 | 2 | 199 | |||
| XM_006231536.2 | XP_006231598.1 | 2 | 196 | |||
| XM_006231537.2 | XP_006231599.1 | 2 | 743 | |||
| GenBank mRNA | BC085125.1 | - | 2 | 90 | 7131883 | |
| CB584165.1 | - | 2 | 174 | |||
| EV771066.1 | - | 2 | 101 | |||

Btrc
Genomic Map