siRNA Details
| Gene Symbol: Stk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013197.2 | NP_001013215.2 | 5 | 602 | ||
| GenBank mRNA | BC061571.1 | - | 5 | 606 | 6921289 | |
| Gene Symbol: LOC103694374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772703.1 | XP_008770925.1 | 606 | |||

Stk19
Genomic Map