siRNA Details
| Gene Symbol: Stk38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015025.1 | NP_001015025.1 | 1, 2 | 297 | ||
| XM_006256139.2 | XP_006256201.1 | 10, 11 | 1237 | |||
| XM_006256140.2 | XP_006256202.1 | 9, 10 | 1156 | |||
| XM_006256141.2 | XP_006256203.1 | 9, 10 | 1203 | |||
| GenBank mRNA | BC091415.1 | - | 1, 2 | 297 | 7365181 | |
| FQ216844.1 | - | 9, 10 | 1086 | |||
| FQ217790.1 | - | 9, 10 | 1086 | |||

Stk38
Genomic Map