siRNA Details
| Gene Symbol: Stk38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015025.1 | NP_001015025.1 | 6 | 757 | ||
| XM_006256139.2 | XP_006256201.1 | 15 | 1697 | |||
| XM_006256140.2 | XP_006256202.1 | 14 | 1616 | |||
| XM_006256141.2 | XP_006256203.1 | 14 | 1663 | |||
| GenBank mRNA | BC091415.1 | - | 6 | 757 | 7365181 | |
| FQ216844.1 | - | 14 | 1546 | |||
| FQ217790.1 | - | 14 | 1546 | |||

Stk38
Genomic Map