siRNA Details
| Gene Symbol: Ybey | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108529.1 | NP_001101999.1 | 2 | 181 | ||
| XM_006256301.2 | XP_006256363.1 | 2 | 192 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ybey| Gene Symbol: Ybey | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108529.1 | NP_001101999.1 | 2 | 181 | ||
| XM_006256301.2 | XP_006256363.1 | 2 | 192 | |||