siRNA Details
| Gene Symbol: Rfesd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108540.1 | NP_001102010.1 | 3 | 422 | ||
| XM_006231706.2 | XP_006231768.1 | 4 | 1522 | |||
| XM_006231708.2 | XP_006231770.1 | 4 | 951 | |||
| XM_006231709.2 | XP_006231771.1 | 4 | 636 | |||

Rfesd
Genomic Map