siRNA Details
| Gene Symbol: Arhgef28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108542.1 | NP_001102012.1 | 32, 33 | 4169 | ||
| XM_006231817.2 | XP_006231879.1 | 24, 25 | 3489 | |||
| XM_008760676.1 | XP_008758898.1 | 31, 32 | 4108 | |||
| XM_008760677.1 | XP_008758899.1 | 31, 32 | 4108 | |||

Arhgef28
Genomic Map