siRNA Details
| Gene Symbol: Arhgef28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108542.1 | NP_001102012.1 | 34, 35 | 4712 | ||
| XM_006231817.2 | XP_006231879.1 | 26, 27 | 4032 | |||
| XM_008760676.1 | XP_008758898.1 | 33, 34 | 4651 | |||
| XM_008760677.1 | XP_008758899.1 | 33, 34 | 4651 | |||

Arhgef28
Genomic Map