siRNA Details
| Gene Symbol: Arhgef28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108542.1 | NP_001102012.1 | 6 | 820 | ||
| XM_008760676.1 | XP_008758898.1 | 5 | 759 | |||
| XM_008760677.1 | XP_008758899.1 | 5 | 759 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef28| Gene Symbol: Arhgef28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108542.1 | NP_001102012.1 | 6 | 820 | ||
| XM_008760676.1 | XP_008758898.1 | 5 | 759 | |||
| XM_008760677.1 | XP_008758899.1 | 5 | 759 | |||