siRNA Details
| Gene Symbol: Bbs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012180.1 | NP_001012180.1 | 8 | 894 | ||
| GenBank mRNA | BC081968.1 | - | 8 | 894 | 7125501 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bbs7| Gene Symbol: Bbs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012180.1 | NP_001012180.1 | 8 | 894 | ||
| GenBank mRNA | BC081968.1 | - | 8 | 894 | 7125501 | |