siRNA Details
Gene Symbol: Spata5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108549.1 | NP_001102019.1 | 8 | 1827 | ||
XM_008760932.1 | XP_008759154.1 | 8 | 1818 | |||
XM_008760933.1 | XP_008759155.1 | 8 | 1477 | |||
XM_008760934.1 | XP_008759156.1 | 8 | 1818 |
