siRNA Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191847.1 | NP_001178776.1 | 6 | 484 | ||
| XM_008761056.1 | XP_008759278.1 | 6 | 486 | |||
| GenBank mRNA | FQ211604.1 | - | 501 | |||
| FQ217493.1 | - | 469 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd1| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191847.1 | NP_001178776.1 | 6 | 484 | ||
| XM_008761056.1 | XP_008759278.1 | 6 | 486 | |||
| GenBank mRNA | FQ211604.1 | - | 501 | |||
| FQ217493.1 | - | 469 | ||||