siRNA Details
| Gene Symbol: Alg14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014176.2 | NP_001014198.1 | 3 | 502 | ||
| XM_006233243.2 | XP_006233305.1 | 3 | 549 | |||
| XM_008761465.1 | XP_008759687.1 | 3 | 549 | |||
| XM_008761466.1 | XP_008759688.1 | 3 | 532 | |||
| GenBank mRNA | BC079151.1 | - | 3 | 448 | 7128610 | |
| CO400199.1 | - | 3 | 502 | |||

Alg14
Genomic Map