siRNA Details
| Gene Symbol: Ap1ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191850.1 | NP_001178779.1 | 10 | 874 | ||
| XM_006233291.2 | XP_006233353.2 | 10 | 1207 | |||
| XM_006233292.2 | XP_006233354.2 | 9 | 1108 | |||
| GenBank mRNA | BC091336.1 | - | 3 | 422 | ||

Ap1ar
Genomic Map