siRNA Details
| Gene Symbol: Ap1ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191850.1 | NP_001178779.1 | 8 | 461 | ||
| XM_006233291.2 | XP_006233353.2 | 8 | 794 | |||
| XM_006233292.2 | XP_006233354.2 | 7 | 695 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap1ar| Gene Symbol: Ap1ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191850.1 | NP_001178779.1 | 8 | 461 | ||
| XM_006233291.2 | XP_006233353.2 | 8 | 794 | |||
| XM_006233292.2 | XP_006233354.2 | 7 | 695 | |||