siRNA Details
| Gene Symbol: Ostc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108566.1 | NP_001102036.1 | 3 | 441 | ||
| GenBank mRNA | BC159425.1 | - | 3 | 427 | 9030828 | |
| FQ219812.1 | - | 3 | 447 | |||
| FQ219920.1 | - | 3 | 455 | |||
| FQ220255.1 | - | 3 | 423 | |||
| FQ230162.1 | - | 3 | 423 | |||
| FQ234814.1 | - | 3 | 403 | |||

Ostc
Genomic Map