siRNA Details
| Gene Symbol: Lamtor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008375.1 | NP_001008376.1 | 2, 3 | 133 | ||
| XM_006233383.2 | XP_006233445.1 | 2, 3 | 258 | |||
| XM_006233385.2 | XP_006233447.1 | 2, 3 | 203 | |||
| GenBank mRNA | BC086353.1 | - | 2, 3 | 133 | 7304027 | |

Lamtor3
Genomic Map