siRNA Details
Gene Symbol: Morn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047912.1 | NP_001041377.1 | 10 | 1037 | ||
GenBank mRNA | AY383696.1 | - | 10 | 1037 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Morn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047912.1 | NP_001041377.1 | 10 | 1037 | ||
GenBank mRNA | AY383696.1 | - | 10 | 1037 |