siRNA Details
| Gene Symbol: Arfgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033707.1 | NP_001028879.1 | 12 | 1201 | ||
| XM_006234529.2 | XP_006234591.1 | 13 | 1375 | |||
| XM_006234530.2 | XP_006234592.1 | 13 | 1347 | |||
| XM_006234531.2 | XP_006234593.1 | 12 | 1332 | |||
| GenBank mRNA | BC101917.1 | - | 12 | 1201 | 7935829 | |
| FQ218185.1 | - | 12 | 1273 | |||
| FQ228393.1 | - | 11 | 2234 | |||

Arfgap2
Genomic Map