siRNA Details
| Gene Symbol: Dnajc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191853.1 | NP_001178782.1 | 3 | 264 | ||
| XM_006234653.2 | XP_006234715.1 | 3 | 308 | |||
| XM_006234654.2 | XP_006234716.1 | 3 | 297 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc24| Gene Symbol: Dnajc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191853.1 | NP_001178782.1 | 3 | 264 | ||
| XM_006234653.2 | XP_006234715.1 | 3 | 308 | |||
| XM_006234654.2 | XP_006234716.1 | 3 | 297 | |||