siRNA Details
| Gene Symbol: Tmem87b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191854.1 | NP_001178783.1 | 11 | 1071 | ||
| XM_008762182.1 | XP_008760404.1 | 11 | 1273 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem87b| Gene Symbol: Tmem87b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191854.1 | NP_001178783.1 | 11 | 1071 | ||
| XM_008762182.1 | XP_008760404.1 | 11 | 1273 | |||