siRNA Details
| Gene Symbol: Lamp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014183.1 | NP_001014205.1 | 2, 3 | 526 | ||
| GenBank mRNA | BC087680.1 | - | 2, 3 | 526 | 7318802 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lamp5| Gene Symbol: Lamp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014183.1 | NP_001014205.1 | 2, 3 | 526 | ||
| GenBank mRNA | BC087680.1 | - | 2, 3 | 526 | 7318802 | |