siRNA Details
Gene Symbol: Dhx35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108601.1 | NP_001102071.1 | 11 | 1020 | ||
XM_006235423.2 | XP_006235485.1 | 11 | 1026 | |||
XM_008762359.1 | XP_008760581.1 | 11 | 1027 | |||
XM_008762360.1 | XP_008760582.1 | 7 | 526 |
