siRNA Details
| Gene Symbol: Dhx35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108601.1 | NP_001102071.1 | 5 | 502 | ||
| XM_006235423.2 | XP_006235485.1 | 5 | 508 | |||
| XM_008762359.1 | XP_008760581.1 | 5 | 509 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhx35| Gene Symbol: Dhx35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108601.1 | NP_001102071.1 | 5 | 502 | ||
| XM_006235423.2 | XP_006235485.1 | 5 | 508 | |||
| XM_008762359.1 | XP_008760581.1 | 5 | 509 | |||