siRNA Details
| Gene Symbol: Cse1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108607.1 | NP_001102077.1 | 23, 24 | 2685 | ||
| XM_006235648.2 | XP_006235710.1 | 24, 25 | 2783 | |||
| GenBank mRNA | FQ233199.1 | - | 4, 5 | 358 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cse1l| Gene Symbol: Cse1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108607.1 | NP_001102077.1 | 23, 24 | 2685 | ||
| XM_006235648.2 | XP_006235710.1 | 24, 25 | 2783 | |||
| GenBank mRNA | FQ233199.1 | - | 4, 5 | 358 | ||