siRNA Details
| Gene Symbol: Sqrdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325246.1 | - | 4 | 323 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sqrdl| Gene Symbol: Sqrdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325246.1 | - | 4 | 323 | ||