siRNA Details
Gene Symbol: Rbm33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191859.1 | NP_001178788.1 | 1672 | |||
XM_006235845.2 | XP_006235907.1 | 3820 | ||||
XM_008762624.1 | XP_008760846.1 | 3940 | ||||
XM_008762625.1 | XP_008760847.1 | 3940 | ||||
XM_008762626.1 | XP_008760848.1 | 3820 | ||||
GenBank mRNA | FQ211558.1 | - | 18 |
