siRNA Details
| Gene Symbol: Mfap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108644.1 | NP_001102114.1 | 9, 10 | 451 | ||
| GenBank mRNA | FQ229611.1 | - | 8, 9 | 557 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfap5| Gene Symbol: Mfap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108644.1 | NP_001102114.1 | 9, 10 | 451 | ||
| GenBank mRNA | FQ229611.1 | - | 8, 9 | 557 | ||