siRNA Details
| Gene Symbol: Clec2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048075.1 | NP_001041540.1 | 4, 5 | 470 | ||
| GenBank mRNA | DQ168417.1 | - | 4, 5 | 470 | ||
| DQ168419.1 | - | 4, 5 | 470 | |||
| EF100690.1 | - | 4, 5 | 470 | |||
| EU128748.1 | - | 4, 5 | 476 | |||

Clec2g
Genomic Map