siRNA Details
| Gene Symbol: Clec2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048075.1 | NP_001041540.1 | 5 | 567 | ||
| GenBank mRNA | DQ168417.1 | - | 5 | 567 | ||
| EF100690.1 | - | 5 | 567 | |||
| EU128748.1 | - | 5 | 573 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec2g| Gene Symbol: Clec2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048075.1 | NP_001041540.1 | 5 | 567 | ||
| GenBank mRNA | DQ168417.1 | - | 5 | 567 | ||
| EF100690.1 | - | 5 | 567 | |||
| EU128748.1 | - | 5 | 573 | |||