siRNA Details
| Gene Symbol: Tox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108654.1 | NP_001102124.1 | 5 | 657 | ||
| XM_006237844.2 | XP_006237906.1 | 5 | 1016 | |||
| XM_008763547.1 | XP_008761769.1 | 4 | 762 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tox| Gene Symbol: Tox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108654.1 | NP_001102124.1 | 5 | 657 | ||
| XM_006237844.2 | XP_006237906.1 | 5 | 1016 | |||
| XM_008763547.1 | XP_008761769.1 | 4 | 762 | |||