siRNA Details
| Gene Symbol: Akna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108668.1 | NP_001102138.1 | 2 | 484 | ||
| XM_006238255.2 | XP_006238317.1 | 1 | 350 | |||
| XM_006238256.2 | XP_006238318.1 | 2 | 829 | |||
| XM_006238257.2 | XP_006238319.1 | 2 | 395 | |||
| XM_008763780.1 | XP_008762002.1 | 2 | 397 | |||
| XM_008763781.1 | XP_008762003.1 | 3 | 2817 | |||
| XM_008763782.1 | XP_008762004.1 | 3 | 2278 | |||
| XM_008763783.1 | XP_008762005.1 | 2 | 1538 | |||
| XM_008763784.1 | XP_008762006.1 | 2 | 2147 | |||

Akna
Genomic Map