siRNA Details
| Gene Symbol: Trit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108676.1 | NP_001102146.1 | 4, 5 | 616 | ||
| XM_006238804.2 | XP_006238866.1 | 4, 5 | 1155 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trit1| Gene Symbol: Trit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108676.1 | NP_001102146.1 | 4, 5 | 616 | ||
| XM_006238804.2 | XP_006238866.1 | 4, 5 | 1155 | |||