siRNA Details
| Gene Symbol: Thap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108695.1 | NP_001102165.1 | 1, 2 | 182 | ||
| GenBank mRNA | BC159427.1 | - | 1, 2 | 80 | 9088591 | |
| FQ229895.1 | - | 1, 2 | 68 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thap3| Gene Symbol: Thap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108695.1 | NP_001102165.1 | 1, 2 | 182 | ||
| GenBank mRNA | BC159427.1 | - | 1, 2 | 80 | 9088591 | |
| FQ229895.1 | - | 1, 2 | 68 | |||