siRNA Details
| Gene Symbol: Srsf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC099175.1 | - | 3 | 496 | 7382701 | |
| FM085512.1 | - | 3 | 500 | |||
| FQ229134.1 | - | 3 | 513 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srsf7| Gene Symbol: Srsf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC099175.1 | - | 3 | 496 | 7382701 | |
| FM085512.1 | - | 3 | 500 | |||
| FQ229134.1 | - | 3 | 513 | |||