siRNA Details
Gene Symbol: Gemin6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009466.2 | NP_001009466.1 | 2 | 181 | ||
GenBank mRNA | AY053516.1 | - | 2 | 182 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gemin6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009466.2 | NP_001009466.1 | 2 | 181 | ||
GenBank mRNA | AY053516.1 | - | 2 | 182 |