siRNA Details
| Gene Symbol: Tssc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012192.1 | NP_001012192.1 | 6 | 652 | ||
| GenBank mRNA | BC087633.1 | - | 6 | 652 | 7320156 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tssc1| Gene Symbol: Tssc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012192.1 | NP_001012192.1 | 6 | 652 | ||
| GenBank mRNA | BC087633.1 | - | 6 | 652 | 7320156 | |