siRNA Details
| Gene Symbol: Coch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108710.1 | NP_001102180.1 | 6 | 515 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coch| Gene Symbol: Coch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108710.1 | NP_001102180.1 | 6 | 515 | ||