siRNA Details
| Gene Symbol: Mbip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108712.2 | NP_001102182.2 | 3 | 407 | ||
| XM_006240120.2 | XP_006240182.1 | 3 | 425 | |||
| GenBank mRNA | BC166982.1 | - | 3 | 334 | ||
| CB609032.1 | - | 3 | 413 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mbip| Gene Symbol: Mbip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108712.2 | NP_001102182.2 | 3 | 407 | ||
| XM_006240120.2 | XP_006240182.1 | 3 | 425 | |||
| GenBank mRNA | BC166982.1 | - | 3 | 334 | ||
| CB609032.1 | - | 3 | 413 | |||