siRNA Details
| Gene Symbol: Ttc7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108719.2 | NP_001102189.1 | 3 | 327 | ||
| XM_008764816.1 | XP_008763038.1 | 9 | 1037 | |||
| GenBank mRNA | BC168996.1 | - | 3 | 327 | 7319639 | |
| FQ210625.1 | - | 5 | 455 | |||

Ttc7b
Genomic Map