siRNA Details
| Gene Symbol: Traf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108724.1 | NP_001102194.1 | 11 | 1322 | ||
| XM_006240596.2 | XP_006240658.1 | 11 | 1367 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Traf3| Gene Symbol: Traf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108724.1 | NP_001102194.1 | 11 | 1322 | ||
| XM_006240596.2 | XP_006240658.1 | 11 | 1367 | |||