siRNA Details
| Gene Symbol: Coq10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108727.1 | NP_001102197.1 | 5 | 601 | ||
| XM_006240759.2 | XP_006240821.1 | 5 | 1770 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coq10a| Gene Symbol: Coq10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108727.1 | NP_001102197.1 | 5 | 601 | ||
| XM_006240759.2 | XP_006240821.1 | 5 | 1770 | |||